COM448632355 Combi-Blocks, Inc.

N-[5-Chloro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetamide

  • MDL number MFCD24039373
  • Cas Number : 1884233-06-0
  • Molecular Weight : 295.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1038

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1038
Availability details for
Quantity
  • Availability
  • Add to Cart