COM448632376 Combi-Blocks, Inc.

2-(2-Methylpropoxy)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

  • MDL number MFCD24039494
  • Molecular Weight : 320.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1060

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1060
Availability details for
Quantity
  • Availability
  • Add to Cart