COM448632385 Combi-Blocks, Inc.

2-Ethoxy-n-(propan-2-yl)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

  • MDL number MFCD24039488
  • Molecular Weight : 333.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1069

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1069
Availability details for
Quantity
  • Availability
  • Add to Cart