COM448632394 Combi-Blocks, Inc.

2-Nitro-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number MFCD27936084
  • Cas Number : 2055777-49-4
  • Molecular Weight : 274.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1078

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1078
Availability details for
Quantity
  • Availability
  • Add to Cart