COM448632411 Combi-Blocks, Inc.

N-Phenyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-1-carboxamide

  • MDL number MFCD24039446
  • Molecular Weight : 373.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1095

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1095
Availability details for
Quantity
  • Availability
  • Add to Cart