COM448632413 Combi-Blocks, Inc.

1-[3-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]cyclopropane-1-carboxamide

  • MDL number MFCD24039917
  • Cas Number : 1628010-77-4
  • Molecular Weight : 287.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1097

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1097
Availability details for
Quantity
  • Availability
  • Add to Cart