COM448632414 Combi-Blocks, Inc.

3-Cyano-3-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propanoic acid

  • MDL number MFCD24040023
  • Molecular Weight : 301.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1098

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1098
Availability details for
Quantity
  • Availability
  • Add to Cart