COM448632415 Combi-Blocks, Inc.

N-tert-Butyl-3-fluoro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

  • MDL number MFCD24039966
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1099

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1099
Availability details for
Quantity
  • Availability
  • Add to Cart