COM448632428 Combi-Blocks, Inc.

4-Formyl-2-methoxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate

  • MDL number MFCD27936099
  • Cas Number : 2130752-10-0
  • Molecular Weight : 320.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1112

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1112
Availability details for
Quantity
  • Availability
  • Add to Cart