COM448632437 Combi-Blocks, Inc.

N-Benzyl-n-methyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

  • MDL number MFCD24039364
  • Molecular Weight : 351.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1121

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1121
Availability details for
Quantity
  • Availability
  • Add to Cart