COM448632449 Combi-Blocks, Inc.

8-(Tetramethyl-1,3,2-dioxaborolan-2-yl)quinolin-5-amine

  • MDL number MFCD27936105
  • Molecular Weight : 270.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1133

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1133
Availability details for
Quantity
  • Availability
  • Add to Cart