COM448632458 Combi-Blocks, Inc.

6-Fluoro-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzothiazol-2-amine

  • MDL number MFCD27936111
  • Molecular Weight : 294.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1142

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1142
Availability details for
Quantity
  • Availability
  • Add to Cart