COM448632460 Combi-Blocks, Inc.

6-Methyl-3-nitro-8-(tetramethyl-1,3,2-dioxaborolan-2-yl)imidazo[1,2-a]pyridine

  • MDL number MFCD27936113
  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1144

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1144
Availability details for
Quantity
  • Availability
  • Add to Cart