COM448632461 Combi-Blocks, Inc.

2,2,2-Trichloro-1-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrol-2-yl]ethanone

  • MDL number MFCD27936114
  • Molecular Weight : 338.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1145

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1145
Availability details for
Quantity
  • Availability
  • Add to Cart