COM448632465 Combi-Blocks, Inc.

5-Methyl-3-(trifluoromethyl)pyrazole-4-boronic acid pinacol ester

  • MDL number MFCD27936117
  • Molecular Weight : 276.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1149

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1149
Availability details for
Quantity
  • Availability
  • Add to Cart