COM448632466 Combi-Blocks, Inc.

7-Methyl-3-nitro-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)imidazo[1,2-a]pyridine

  • MDL number MFCD27936118
  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1150
Availability details for
Quantity
  • Availability
  • Add to Cart