COM448632472 Combi-Blocks, Inc.

Methyl 4-chloro-8-(tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline-2-carboxylate

  • MDL number MFCD27936124
  • Molecular Weight : 347.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1156

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1156
Availability details for
Quantity
  • Availability
  • Add to Cart