COM448632473 Combi-Blocks, Inc.

3-(4-Methylphenyl)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-pyrazole

  • MDL number MFCD27936125
  • Molecular Weight : 284.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1157

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1157
Availability details for
Quantity
  • Availability
  • Add to Cart