COM448632475 Combi-Blocks, Inc.

Methyl 5-fluoro-4-hydroxy-8-(tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline-2-carboxylate

  • MDL number MFCD27936127
  • Molecular Weight : 347.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1159

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1159
Availability details for
Quantity
  • Availability
  • Add to Cart