COM448632479 Combi-Blocks, Inc.

3-(4-Fluorophenyl)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-pyrazole

  • MDL number MFCD27936131
  • Molecular Weight : 288.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1163

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1163
Availability details for
Quantity
  • Availability
  • Add to Cart