COM448632490 Combi-Blocks, Inc.

Methyl 5-(2-chlorophenyl)-1-[5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]pyrazole-4-carboxylate

  • MDL number MFCD27936140
  • Molecular Weight : 439.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1174

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1174
Availability details for
Quantity
  • Availability
  • Add to Cart