COM448632493 Combi-Blocks, Inc.

Methyl 6-chloro-4-oxo-8-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,4-dihydroquinoline-2-carboxylate

  • MDL number MFCD24039375
  • Molecular Weight : 363.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1177

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1177
Availability details for
Quantity
  • Availability
  • Add to Cart