COM448632495 Combi-Blocks, Inc.

2,2-Difluoro-2-[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethoxy)phenyl]acetic acid

  • MDL number MFCD24039974
  • Molecular Weight : 382.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1179

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1179
Availability details for
Quantity
  • Availability
  • Add to Cart