COM448632497 Combi-Blocks, Inc.

N-(3-methoxypropyl)-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

  • MDL number MFCD18734945
  • Cas Number : 1421326-03-5
  • Molecular Weight : 292.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1181

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1181
Availability details for
Quantity
  • Availability
  • Add to Cart