COM448632528 Combi-Blocks, Inc.

6-Methoxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzothiazole-2-thiol

  • MDL number MFCD27936165
  • Molecular Weight : 323.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1216

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1216
Availability details for
Quantity
  • Availability
  • Add to Cart