COM448632532 Combi-Blocks, Inc.

4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)-1,3-benzothiazole-2-thiol

  • MDL number MFCD27936167
  • Molecular Weight : 361.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Freezer

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1220
Availability details for
Quantity
  • Availability
  • Add to Cart