COM448632536 Combi-Blocks, Inc.

2-Hydroxy-2-methyl-n-[6-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-yl]propanamide

  • MDL number MFCD24039690
  • Cas Number : 2377607-33-3
  • Molecular Weight : 355.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1225

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1225
Availability details for
Quantity
  • Availability
  • Add to Cart