COM448632537 Combi-Blocks, Inc.

2-Methyl-n-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2-propenamide

  • MDL number MFCD22494183
  • Cas Number : 1056904-41-6
  • Molecular Weight : 287.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1227
Availability details for
Quantity
  • Availability
  • Add to Cart