COM448632539 Combi-Blocks, Inc.

6-Chloro-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-2h-1,2,3-benzotriazole

  • MDL number MFCD24039759
  • Molecular Weight : 279.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Freezer

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1229
Availability details for
Quantity
  • Availability
  • Add to Cart