COM448632544 Combi-Blocks, Inc.

5-Fluoro-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)-3H-1,2,3-benzotriazole

  • MDL number MFCD27936172
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1234
Availability details for
Quantity
  • Availability
  • Add to Cart