COM448632548 Combi-Blocks, Inc.

1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidin-2-one

  • MDL number MFCD22415355
  • Cas Number : 1007346-42-0
  • Molecular Weight : 301.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1241

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1241
Availability details for
Quantity
  • Availability
  • Add to Cart