COM448632549 Combi-Blocks, Inc.

1-[2-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)phenyl]piperidin-2-one

  • MDL number MFCD24039706
  • Molecular Weight : 369.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1242

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1242
Availability details for
Quantity
  • Availability
  • Add to Cart