COM448632569 Combi-Blocks, Inc.

Benzyl n-[2-[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-yl]carbamate

  • MDL number MFCD24039348
  • Cas Number : 887254-71-9
  • Molecular Weight : 395.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1262

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1262
Availability details for
Quantity
  • Availability
  • Add to Cart