COM448632570 Combi-Blocks, Inc.

Benzyl n-[2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-yl]carbamate

  • MDL number MFCD24039730
  • Cas Number : 850197-54-5
  • Molecular Weight : 395.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1263

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1263
Availability details for
Quantity
  • Availability
  • Add to Cart