COM448632596 Combi-Blocks, Inc.

1,3-Dimethyl-8-(tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxalin-2-one

  • MDL number MFCD27936207
  • Molecular Weight : 300.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1289

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1289
Availability details for
Quantity
  • Availability
  • Add to Cart