COM448632606 Combi-Blocks, Inc.

N-Hexyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonamide

  • MDL number MFCD27936217
  • Molecular Weight : 367.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1299

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1299
Availability details for
Quantity
  • Availability
  • Add to Cart