COM448632618 Combi-Blocks, Inc.

N-(Furan-2-ylmethyl)-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzenesulfonamide

  • MDL number MFCD27936229
  • Molecular Weight : 431.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1311

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1311
Availability details for
Quantity
  • Availability
  • Add to Cart