COM448632624 Combi-Blocks, Inc.

2-Amino-4-methoxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile

  • MDL number MFCD27936235
  • Molecular Weight : 275.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1317

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1317
Availability details for
Quantity
  • Availability
  • Add to Cart