COM448632625 Combi-Blocks, Inc.

N-Ethyl-3-methyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonamide

  • MDL number MFCD27936236
  • Molecular Weight : 325.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1318

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1318
Availability details for
Quantity
  • Availability
  • Add to Cart