COM448632627 Combi-Blocks, Inc.

N-Ethyl-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine hydrochloride

  • MDL number MFCD27936238
  • Molecular Weight : 284.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1320

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1320
Availability details for
Quantity
  • Availability
  • Add to Cart