COM448632634 Combi-Blocks, Inc.

4-Hydroxy-8-(tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline-3-carboxylic acid

  • MDL number MFCD27936244
  • Molecular Weight : 315.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1327

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1327
Availability details for
Quantity
  • Availability
  • Add to Cart