COM448632637 Combi-Blocks, Inc.

N-Ethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethoxy)benzenesulfonamide

  • MDL number MFCD27936246
  • Molecular Weight : 395.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1330

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1330
Availability details for
Quantity
  • Availability
  • Add to Cart