COM448632642 Combi-Blocks, Inc.

N-tert-Butyl-3-methyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonamide

  • MDL number MFCD27936251
  • Molecular Weight : 353.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1335

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1335
Availability details for
Quantity
  • Availability
  • Add to Cart