COM448632644 Combi-Blocks, Inc.

3-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4,5-dihydro-1,2,4-oxadiazol-5-one

  • MDL number MFCD24039940
  • Cas Number : 1454814-36-8
  • Molecular Weight : 288.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1337

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1337
Availability details for
Quantity
  • Availability
  • Add to Cart