COM448632647 Combi-Blocks, Inc.

4-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2H-1,2,4-triazol-3-one

  • MDL number MFCD27936255
  • Cas Number : 2377608-66-5
  • Molecular Weight : 287.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1340
Availability details for
Quantity
  • Availability
  • Add to Cart