COM448632654 Combi-Blocks, Inc.

3-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4h-1,2,4-triazole

  • MDL number MFCD24039958
  • Cas Number : 2377610-67-6
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1347

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1347
Availability details for
Quantity
  • Availability
  • Add to Cart