COM448632657 Combi-Blocks, Inc.

Methyl 8-methyl-4-oxo-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-quinoline-2-carboxylate

  • MDL number MFCD27936264
  • Molecular Weight : 343.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1350

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1350
Availability details for
Quantity
  • Availability
  • Add to Cart