COM448632658 Combi-Blocks, Inc.

6-chloro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine

  • MDL number MFCD18723301
  • Molecular Weight : 254.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1351

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1351
Availability details for
Quantity
  • Availability
  • Add to Cart