COM448632659 Combi-Blocks, Inc.

Methyl 4-oxo-8-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,4-dihydroquinoline-2-carboxylate

  • MDL number MFCD24039376
  • Molecular Weight : 329.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1352

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1352
Availability details for
Quantity
  • Availability
  • Add to Cart