COM448632669 Combi-Blocks, Inc.

2-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD27936273
  • Molecular Weight : 239.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1365

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1365
Availability details for
Quantity
  • Availability
  • Add to Cart